Thermo Scientific Chemicals

Azelaic acid, 98%, Thermo Scientific Chemicals

Catalog number: 036308.22
100 g, Each
Thermo Scientific Chemicals

Azelaic acid, 98%, Thermo Scientific Chemicals

Catalog number: 036308.22
100 g, Each
Quantity
Catalog number: 036308.22
also known as 036308-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-99-9
IUPAC Name
nonanedioic acid
Molecular Formula
C9H16O4
InChI Key
BDJRBEYXGGNYIS-UHFFFAOYSA-N
SMILES
OC(=O)CCCCCCCC(O)=O
Appearance (Color)
White to yellow
Assay (Titration)
>97.5%
Form
Powder or flakes

Description

Useful in lubrication and plasticizers. Azelaic acid is used to treat acne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text