Thermo Scientific Chemicals

Lead(II) telluride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 036317.18
Each
Thermo Scientific Chemicals

Lead(II) telluride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 036317.18
Each
Catalog number: 036317.18
also known as 036317-18
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1314-91-6
Total Metal Impurities
0.01% max.

Description

In semiconductor research, photoconductor cellsLead(II) telluride is used as an infrared detector material. It is also used in spacecraft power applications. Further, it is used in infrared detection, imaging and in solar cells. In addition, it acts as a semiconductor and photoconductor in the form of single crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Resistant to acids
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text