Thermo Scientific Chemicals

Zirconium silicide, 99.5% (metals basis excluding Hf), Hf 4% max, Thermo Scientific Chemicals

Catalog number: 036322.22
100 g, Each
Thermo Scientific Chemicals

Zirconium silicide, 99.5% (metals basis excluding Hf), Hf 4% max, Thermo Scientific Chemicals

Catalog number: 036322.22
100 g, Each
Quantity
Catalog number: 036322.22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12039-90-6
Form
Powder
Assay from Supplier's CofA
≥99.5% (metals basis)
Particle Size
-325 mesh (≤44 microns)
Appearance (Color)
Grey

Description

Zirconium silicate is used for manufacturing refractory materials. It is also used in production of some ceramics, enamels, and ceramic glazes. In enamels and glazes it serves as an opacifier. zirconium silicate is as beads for milling and grinding.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text