Aluminum Fluoride, Anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum Fluoride, Anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum Fluoride, Anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals

For vacuum deposition
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 036335.06
also known as 036335-06
Price (USD)/ Each
84.65
Online exclusive
94.30 
Save 9.65 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
84.65
Online exclusive
94.30 
Save 9.65 (10%)
Add to cart
Aluminum Fluoride, Anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number036335.06
Price (USD)/ Each
84.65
Online exclusive
94.30 
Save 9.65 (10%)
-
Add to cart
Chemical Identifiers
CAS7784-18-1
IUPAC Namealuminium(3+) trifluoride
Molecular FormulaAlF3
InChI KeyKLZUFWVZNOTSEM-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey
FormPieces or lumps, 3-12mm approximate
Total Metal Impurities≤0.5%
Aluminum fluoride (AlF3) is an inorganic compound used primarily in the production of aluminum. It is also used to inhibit fermentation. It is a sputtering target for preparation of low index films. In ceramics, as flux in metallurgy, in aluminum manufacture, as inhibitor of fermentation, as catalyst in organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only