Thermo Scientific Chemicals

Aluminum Fluoride, Anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 036335.14
25 g, Each
Thermo Scientific Chemicals

Aluminum Fluoride, Anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 036335.14
25 g, Each
Quantity
Catalog number: 036335.14
also known as 036335-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7784-18-1
IUPAC Name
aluminium(3+) trifluoride
Molecular Formula
AlF3
InChI Key
KLZUFWVZNOTSEM-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[Al+3]
Appearance (Color)
White to grey
Form
Pieces or lumps, 3-12mm approximate
Total Metal Impurities
≤0.5%

Description

Aluminum fluoride (AlF3) is an inorganic compound used primarily in the production of aluminum. It is also used to inhibit fermentation. It is a sputtering target for preparation of low index films. In ceramics, as flux in metallurgy, in aluminum manufacture, as inhibitor of fermentation, as catalyst in organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text