Thermo Scientific Chemicals

Nickel(II) sulfate hexahydrate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 036336.22
100 g, Each
Thermo Scientific Chemicals

Nickel(II) sulfate hexahydrate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 036336.22
100 g, Each
Quantity
Catalog number: 036336.22
also known as 036336-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10101-97-0
IUPAC Name
nickel(2+) hexahydrate sulfate
Molecular Formula
H12NiO10S
InChI Key
RRIWRJBSCGCBID-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
Impurity content
Co 0.002%
Impurity content
Cu 0.005%
Impurity content
Fe 0.001%
Impurity content
K 0.01%
Impurity content
Mg 0.005%

Description

It is the precursor material for Ni electroplating. It is applied in biochemistry and also employed as a calibrant for magnetic susceptibility measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text