Thermo Scientific Chemicals

Butyl stearate, tech., Thermo Scientific Chemicals

Catalog number: 036338.A3
2 kg, Each
Thermo Scientific Chemicals

Butyl stearate, tech., Thermo Scientific Chemicals

Catalog number: 036338.A3
2 kg, Each
Quantity
Catalog number: 036338.A3
also known as 036338-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-95-5
IUPAC Name
butyl octadecanoate
Molecular Formula
C22H44O2
InChI Key
ULBTUVJTXULMLP-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCC(=O)OCCCC
Iodine value
Saponification Value
165 to 180

Description

Butyl stearate is used as finishing agents, lubricants and lubricant additives. It is also used as a plasticizer, food packaging material and as a dye solvent. It acts as a reactant and involved in the preparation of octadecanoic acid methyl ester by reacting with methanol. It finds application as a binder in cosmetics like soaps, shampoos and shaving creams, skin conditioners and surfactants for cosmetic formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text