Indium tin oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Indium tin oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indium tin oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 036348.14
also known as 036348-14
Price (USD)/ Each
108.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
108.00
Add to cart
Indium tin oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number036348.14
Price (USD)/ Each
108.00
-
Add to cart
Chemical Identifiers
CAS50926-11-9
IUPAC Namestannanone indium
Molecular FormulaInOSn
InChI KeyAMGQUBHHOARCQH-UHFFFAOYSA-N
SMILES[In].O=[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Indium tin oxide is used in nanotechnology to provide a path to a new generation of solar cells, to make transparent conductive coatings for displays such as liquid crystal displays, flat panel displays, plasma displays, touch panels, and electronic ink. Thin films of ITO are also used in organic light-emitting diodes, antistatic coatings and EMI shielding. In organic light-emitting diodes, ITO is used as the anode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only