Gadolinium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals
Gadolinium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gadolinium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 036355.14
also known as 036355-14
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
Add to cart
Gadolinium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals
Catalog number036355.14
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS12064-62-9
IUPAC Namedigadolinium(3+) trioxidandiide
Molecular FormulaGd2O3
InChI KeyCMIHHWBVHJVIGI-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Gd+3].[Gd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Gadolinium(III) oxide is used for vacuum deposition, synthesis of raw material for intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only