Holmium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals
Holmium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Holmium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 036357.09
also known as 036357-09
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
Add to cart
Holmium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals
Catalog number036357.09
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS12055-62-8
IUPAC Namediholmium(3+) trioxidandiide
Molecular FormulaHo2O3
InChI KeyOWCYYNSBGXMRQN-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Ho+3].[Ho+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only