Magnesium acetate tetrahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Magnesium acetate tetrahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium acetate tetrahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 036358.22
also known as 036358-22
Price (USD)/ Each
31.65
Online exclusive
34.80 
Save 3.15 (9%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
31.65
Online exclusive
34.80 
Save 3.15 (9%)
Add to cart
Magnesium acetate tetrahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Catalog number036358.22
Price (USD)/ Each
31.65
Online exclusive
34.80 
Save 3.15 (9%)
-
Add to cart
Chemical Identifiers
CAS16674-78-5
IUPAC Namemagnesium(2+) diacetate tetrahydrate
Molecular FormulaC4H14MgO8
InChI KeyXKPKPGCRSHFTKM-UHFFFAOYSA-L
SMILESO.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.0-102.0% C₄H₆MgO₄.4H₂O
Impurity contentBa 0.001%
Impurity contentCa 0.01%
Impurity contentCl 0.001%
Impurity contentFe 5ppm
View more
Used as a source of magnesium in biological reactions and as a source of acetate ion in chemical transformations. Acts as an emission control agent in coal combustion process to reduce acid rain. It is involved in glass formation process via the thermal decomposition of solids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only