D-(-)-Isoascorbic acid, 98%, Thermo Scientific Chemicals
D-(-)-Isoascorbic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-(-)-Isoascorbic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 036366.22
also known as 036366-22
Price (USD)/ Each
35.65
Online exclusive
39.60 
Save 3.95 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
35.65
Online exclusive
39.60 
Save 3.95 (10%)
Add to cart
D-(-)-Isoascorbic acid, 98%, Thermo Scientific Chemicals
Catalog number036366.22
Price (USD)/ Each
35.65
Online exclusive
39.60 
Save 3.95 (10%)
-
Add to cart
Chemical Identifiers
CAS89-65-6
SpecificationsSpecification SheetSpecification Sheet
Formcrystals
Appearance (Color)White to pale yellow
Assay (Titration)> 98.5%
Optical Rotation-16.0 to -19.0 deg ( c=10, water)
D-(-)-Isoascorbic acid is used as antioxidant especially in brewing industry, reducing agent in photography. And it is also used in food industry, as food additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only