Thermo Scientific Chemicals

Calcium fluoride, Vacuum Deposition Grade, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 036377.09
10 g, Each
Thermo Scientific Chemicals

Calcium fluoride, Vacuum Deposition Grade, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 036377.09
10 g, Each
Quantity
Catalog number: 036377.09
also known as 036377-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-75-5
IUPAC Name
calcium difluoride
Molecular Formula
CaF2
InChI Key
WUKWITHWXAAZEY-UHFFFAOYSA-L
SMILES
[F-].[F-].[Ca++]
Total Metal Impurities
0.1% max.

Description

Calcium fluoride is used to produce a wide range of materials, manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text