Barium fluoride, 99.9% (metals basis), Thermo Scientific Chemicals
Barium fluoride, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium fluoride, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 036380.22
also known as 036380-22
Price (USD)/ Each
457.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
457.00
Add to cart
Barium fluoride, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number036380.22
Price (USD)/ Each
457.00
-
Add to cart
Chemical Identifiers
CAS7787-32-8
IUPAC Namebarium(2+) difluoride
Molecular FormulaBaF2
InChI KeyOYLGJCQECKOTOL-UHFFFAOYSA-L
SMILES[F-].[F-].[Ba++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless or white
FormCrystals or powder or crystalline powder
Total Metal Impurities≤0.1% (≤1,000ppm)
Barium fluoride is used as a material to make optical components such as lenses, in windows for infrared spectroscopy, in particular in the field of fuel oil analysis, used as a preopacifying agent and in enamel and glazing frits production. It is also used in the production of welding agents, metallurgy, as a molten bath for refining aluminum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only