Germanium(IV) oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Germanium(IV) oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Germanium(IV) oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Used in Phosphors, Transistors and Diodes, and Infrared Transmitting Glass
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 036382.04
also known as 036382-04
Price (USD)/ Each
64.10
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
64.10
Add to cart
Germanium(IV) oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number036382.04
Price (USD)/ Each
64.10
-
Add to cart
Chemical Identifiers
CAS1310-53-8
IUPAC Namegermanedione
Molecular FormulaGeO2
InChI KeyYBMRDBCBODYGJE-UHFFFAOYSA-N
SMILESO=[Ge]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Used in phosphors, transistors and diodes, and infrared transmitting glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only