Thermo Scientific Chemicals

Magnesium fluoride, 99.9% (metals basis excluding Ca & Na), Ca+Na <1%, Thermo Scientific Chemicals

Catalog number: 036391.14
25 g, Each
Thermo Scientific Chemicals

Magnesium fluoride, 99.9% (metals basis excluding Ca & Na), Ca+Na <1%, Thermo Scientific Chemicals

Catalog number: 036391.14
25 g, Each
Quantity
Catalog number: 036391.14
also known as 036391-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-40-6
IUPAC Name
magnesium(2+) difluoride
Molecular Formula
F2Mg
InChI Key
ORUIBWPALBXDOA-UHFFFAOYSA-L
SMILES
[F-].[F-].[Mg++]
Total Metal Impurities
0.1% max.

Description

Magnesium fluoride is used as Vacuum deposition, anti-reflective coatings, in optics and also used in space telescopes, glass industry and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text