Lead(II) acetate, basic, ACS, Thermo Scientific Chemicals
Lead(II) acetate, basic, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) acetate, basic, ACS, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036393.36
also known as 036393-36
Price (USD)/ Each
78.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
92.00 
Save 13.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
78.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
92.00 
Save 13.35 (15%)
Add to cart
Lead(II) acetate, basic, ACS, Thermo Scientific Chemicals
Catalog number036393.36
Price (USD)/ Each
78.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
92.00 
Save 13.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1335-32-6
IUPAC Nametris(λ²-lead(2+)) diacetate tetrahydroxide
Molecular FormulaC4H10O8Pb3
InChI KeyVLOJXAQYHIVPFI-UHFFFAOYSA-H
SMILES[OH-].[OH-].[OH-].[OH-].[Pb++].[Pb++].[Pb++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA99.0-103.0%
Impurity contentChloride (Cl): ≤5 ppm
Impurity contentCopper (Cu): ≤0.002%
Impurity contentIron (Fe): ≤0.001%
Impurity contentNitrate and Nitrite (as NO₃): ≤0.005%
View more
Lead(II) acetate is used in the production of sweeteners, cosmetics, astringent, used in the cleaning and maintenance of stainless steel firearm suppressors (silencers) and compensators, and also used in making of slow matches.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only