Calcium carbonate, chelometric standard, ACS, 99.95-100.05%, Thermo Scientific Chemicals
Calcium carbonate, chelometric standard, ACS, 99.95-100.05%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium carbonate, chelometric standard, ACS, 99.95-100.05%, Thermo Scientific Chemicals

Calcium carbonate, CCaO3, CAS Number-471-34-1, aeromatt, akadama, aragonite, calcite, calcium carbonate, calofort u, carbonic acid calcium salt 1:1, chalk, limestone, marble, 500g, White powder, 99.95-100.05%, CHEBI:3311, 2.93, 207-439-9, 100.09, ACS, VTYYLEPIZMXCLO-UHFFFAOYSA-L, calcium;carbonate
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 036399.22
also known as 036399-22
Price (USD)/ Each
59.65
Online exclusive
66.00 
Save 6.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
59.65
Online exclusive
66.00 
Save 6.35 (10%)
Add to cart
Calcium carbonate, chelometric standard, ACS, 99.95-100.05%, Thermo Scientific Chemicals
Catalog number036399.22
Price (USD)/ Each
59.65
Online exclusive
66.00 
Save 6.35 (10%)
-
Add to cart
Chemical Identifiers
CAS471-34-1
IUPAC Namecalcium carbonate
Molecular FormulaCCaO3
InChI KeyVTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES[Ca++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.95-100.05% CaCO₃
Heavy Metals(ICP-OES) 0.001%
Impurity contentBa 0.01%
Impurity contentCl 0.001%
Impurity contentF 0.0015%
View more
Calcium carbonate is a dietary supplement used when the quantity of calcium taken in the diet is not enough. Used as an antacid to relieve heartburn, acid indigestion and upset belly. Practiced in the expression industry. Also widely used as a filler in plastics and is used in metallurgy of iron.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only