Thermo Scientific Chemicals

Hydrogen tetrachloroaurate(III) trihydrate, ACS, 99.99% (metals basis), Au 49.0% min, Thermo Scientific Chemicals

Catalog number: 036400.03
1 g, Each
Thermo Scientific Chemicals

Hydrogen tetrachloroaurate(III) trihydrate, ACS, 99.99% (metals basis), Au 49.0% min, Thermo Scientific Chemicals

Catalog number: 036400.03
1 g, Each
Quantity
Catalog number: 036400.03
also known as 036400-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16961-25-4
IUPAC Name
hydrogen tetrachlorogolduide
Molecular Formula
AuCl4H5
InChI Key
QJXOQVKCBDMTLU-UHFFFAOYSA-O
SMILES
[H+].Cl.Cl.Cl.Cl.[Au-]
Assay (unspecified)
49.0% Au min
Impurity content
Alkalies and other metals (as sulfates) 0.2%
Impurity content
Insoluble in ether 0.1%
Total Metal Impurities
0.01% max.

Description

It is used for microanalysis of rubidium, cesium and in the determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in the control rods of nuclear reactors and in the manufacturing of electric light filaments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text