Thermo Scientific Chemicals

Chlorobenzene, ACS, 99.5%, Thermo Scientific Chemicals

Catalog number: 036401.M1
4 x 1 L, Each
Thermo Scientific Chemicals

Chlorobenzene, ACS, 99.5%, Thermo Scientific Chemicals

Catalog number: 036401.M1
4 x 1 L, Each
Quantity
Catalog number: 036401.M1
also known as 036401-M1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-90-7
IUPAC Name
chlorobenzene
Molecular Formula
C6H5Cl
InChI Key
MVPPADPHJFYWMZ-UHFFFAOYSA-N
SMILES
ClC1=CC=CC=C1
Color (APHA)
≤30
Assay from Supplier's CofA
≥99.5%
Free acid (titration)
≤0.004meq/g
Residue on Evaporation
≤0.02%

Description

Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text