Thermo Scientific Chemicals

L-(+)-Tartaric acid, ACS, Thermo Scientific Chemicals

Catalog number: 036405.22
100 g, Each
Thermo Scientific Chemicals

L-(+)-Tartaric acid, ACS, Thermo Scientific Chemicals

Catalog number: 036405.22
100 g, Each
Quantity
Catalog number: 036405.22
also known as 036405-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87-69-4
IUPAC Name
2,3-dihydroxybutanedioic acid
Molecular Formula
C4H6O6
InChI Key
FEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILES
OC(C(O)C(O)=O)C(O)=O
Residue on ignition
0.02%
Impurity content
C₂O₄ P.T. (limit about 0.1%)
Impurity content
Cl 0.001%
Impurity content
Fe 5ppm
Impurity content
PO₄ 0.001%

Description

L-(+)-Tartaric acid is widely utilized in pharmaceutical industries. It is used in soft drinks, confectionaries, food products, gelatin desserts and buffering agents. It forms a compound, TiCl2(O-i-Pr)2 with Diels-Alder catalyst and act as a chelate agent in metal industries. Owing to its efficient chelating property towards metal ions, it is used in the farming and metal industries for complexing micronutrients and for cleaning metal surfaces, respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text