Thermo Scientific Chemicals

Lactic acid, ACS, 85.0-90.0% aq. soln., Thermo Scientific Chemicals

Catalog number: 036415.22
100 g, Each
Thermo Scientific Chemicals

Lactic acid, ACS, 85.0-90.0% aq. soln., Thermo Scientific Chemicals

Catalog number: 036415.22
100 g, Each
Quantity
Catalog number: 036415.22
also known as 036415-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-21-5
IUPAC Name
2-hydroxypropanoic acid
Molecular Formula
C3H6O3
InChI Key
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
SMILES
CC(O)C(O)=O
Appearance (Color)
Clear colorless
Form
Syrupy liquid
Assay from Suppliers CofA
85.0-90.0%
Chloride content
≤0.001%
Residue on ignition
≤0.02%

Description

It is used most commonly by athletes to describe the intense pain felt during exhaustive exercise. Used to make cultured dairy products, as a food preservative, and to make chemicals. Useful for the creation of highly aligned electrospun poly-L-lactic acid fibers for nerve regeneration applications. Poly(lactic acid) nanocomposites with various organoclays shows useful application for gas permeability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text