Hydroxylamine hydrochloride, ACS, 96+%, Thermo Scientific Chemicals
Hydroxylamine hydrochloride, ACS, 96+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydroxylamine hydrochloride, ACS, 96+%, Thermo Scientific Chemicals

A Monoamine Oxidase Inhibitor; Inhibits Platelet Aggregation
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 036416.22
also known as 036416-22
Price (USD)/ Each
48.65
Online exclusive
54.10 
Save 5.45 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
48.65
Online exclusive
54.10 
Save 5.45 (10%)
Add to cart
Hydroxylamine hydrochloride, ACS, 96+%, Thermo Scientific Chemicals
Catalog number036416.22
Price (USD)/ Each
48.65
Online exclusive
54.10 
Save 5.45 (10%)
-
Add to cart
Chemical Identifiers
CAS5470-11-1
IUPAC Namehydroxylamine hydrochloride
Molecular FormulaClH4NO
InChI KeyWTDHULULXKLSOZ-UHFFFAOYSA-N
SMILESCl.NO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals
Assay from Supplier's CofA≥96.0% H₃NO.HCl (Titration of the reductive capacity)
ClarityAlcohol solution: Passes test
Free acid (titration)0.25 meq/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only