Thermo Scientific Chemicals

3,4,5-Trihydroxybenzoic acid monohydrate, ACS, 98+%, Thermo Scientific Chemicals

Catalog number: 036417.22
100 g, Each
Thermo Scientific Chemicals

3,4,5-Trihydroxybenzoic acid monohydrate, ACS, 98+%, Thermo Scientific Chemicals

Catalog number: 036417.22
100 g, Each
Quantity
Catalog number: 036417.22
also known as 036417-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5995-86-8
IUPAC Name
3,4,5-trihydroxybenzoate
Molecular Formula
C7H5O5
InChI Key
LNTHITQWFMADLM-UHFFFAOYSA-M
SMILES
OC1=CC(=CC(O)=C1O)C([O-])=O
Residue on ignition
0.05%
Impurity content
SO₄ 0.02%
Insoluble material
0.01%

Description

3,4,5-Trihydroxybenzoic acid monohydrate is used in making dyes and inks, photographic developers and has been used as astringents in medically. It is also are used as antioxidants in foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text