Thermo Scientific Chemicals

Cobalt(II) nitrate hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 036418.22
100 g, Each
Thermo Scientific Chemicals

Cobalt(II) nitrate hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 036418.22
100 g, Each
Quantity
Catalog number: 036418.22
also known as 036418-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10026-22-9
IUPAC Name
λ²-cobalt(2+) hexahydrate dinitrate
Molecular Formula
CoH12N2O12
InChI Key
QGUAJWGNOXCYJF-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Assay (unspecified)
98.0-102.0% Co(NO₃)₂.6H₂O
Impurity content
Ca 0.005%
Impurity content
Cl 0.002%
Impurity content
Cu 0.002%
Impurity content
Fe 0.001%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications:
  • As battery additive, in manufacture of cobalt pigments and invisible inks, for decorating stoneware and procelain, and in preparation of catalysts
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text