Cobalt(II) nitrate hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Cobalt(II) nitrate hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) nitrate hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

For Use As a Battery Additive, in the Manufacture of Cobalt Pigments and Invisible Inks, For Decorating Stoneware and Porcelain
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Catalog number 036418.36
also known as 036418-36
Price (USD)/ Each
218.65
Online exclusive
243.00 
Save 24.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
218.65
Online exclusive
243.00 
Save 24.35 (10%)
Add to cart
Cobalt(II) nitrate hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Catalog number036418.36
Price (USD)/ Each
218.65
Online exclusive
243.00 
Save 24.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10026-22-9
IUPAC Nameλ²-cobalt(2+) hexahydrate dinitrate
Molecular FormulaCoH12N2O12
InChI KeyQGUAJWGNOXCYJF-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentSO₄ 0.005%
Impurity contentZn 0.01%
Insoluble material0.01%
Assay (unspecified)98.0-102.0% Co(NO₃)₂.6H₂O
Impurity contentCa 0.005%
View more

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications:
  • As battery additive, in manufacture of cobalt pigments and invisible inks, for decorating stoneware and procelain, and in preparation of catalysts
RUO – Research Use Only