Acetonitrile, ACS, 99.5+%, Thermo Scientific Chemicals
Acetonitrile, ACS, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetonitrile, ACS, 99.5+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 x 1 L
4 L
4 x 4 L
Catalog number 036423.AP
also known as 036423-AP
Price (USD)/ Each
48.65
Online exclusive
54.00 
Save 5.35 (10%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
48.65
Online exclusive
54.00 
Save 5.35 (10%)
Add to cart
Acetonitrile, ACS, 99.5+%, Thermo Scientific Chemicals
Catalog number036423.AP
Price (USD)/ Each
48.65
Online exclusive
54.00 
Save 5.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water≤0.3%
FormLiquid
Free base (titration)≤0.0006meq/g
Residue on Evaporation≤0.005%
Assay (unspecified)≥99.5%
View more
It is utilized as a polar aprotic solvent in organic synthesis and in the purification of butadiene. It is used as a mobile phase in HPLC and in LC-MS. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only