Thermo Scientific Chemicals

Acetonitrile, ACS, 99.5+%, Thermo Scientific Chemicals

Catalog number: 036423.M6
4 x 4 L, Each
Thermo Scientific Chemicals

Acetonitrile, ACS, 99.5+%, Thermo Scientific Chemicals

Catalog number: 036423.M6
4 x 4 L, Each
Quantity
Catalog number: 036423.M6
also known as 036423-M6
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-05-8
IUPAC Name
acetonitrile
Molecular Formula
C2H3N
InChI Key
WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES
CC#N
Residual water
≤0.3%
Form
Liquid
Free base (titration)
≤0.0006meq/g
Residue on Evaporation
≤0.005%
Assay (unspecified)
≥99.5%

Description

It is utilized as a polar aprotic solvent in organic synthesis and in the purification of butadiene. It is used as a mobile phase in HPLC and in LC-MS. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text