Thermo Scientific Chemicals

Urea, ACS, 99.0-100.5%, Thermo Scientific Chemicals

Catalog number: 036428.22
100 g, Each
Thermo Scientific Chemicals

Urea, ACS, 99.0-100.5%, Thermo Scientific Chemicals

Catalog number: 036428.22
100 g, Each
Quantity
Catalog number: 036428.22
also known as 036428-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57-13-6
IUPAC Name
urea
Molecular Formula
CH4N2O
InChI Key
XSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILES
NC(N)=O
Melting Point
132-135?C
Assay (unspecified)
99.0-100.5%
Chloride content
≤ 5ppm
Residue on ignition
≤ 0.01%
Impurity content
Sulfate SO₄: 0.001% max

Description

Urea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text