Urea, ACS, 99.0-100.5%, Thermo Scientific Chemicals
Urea, ACS, 99.0-100.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Urea, ACS, 99.0-100.5%, Thermo Scientific Chemicals

Used as a protein denaturant
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
100 g
500 g
Catalog number 036428.A3
also known as 036428-A3
Price (USD)/ Each
79.65
Online exclusive
88.10 
Save 8.45 (10%)
-
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (USD)/ Each
79.65
Online exclusive
88.10 
Save 8.45 (10%)
Add to cart
Urea, ACS, 99.0-100.5%, Thermo Scientific Chemicals
Catalog number036428.A3
Price (USD)/ Each
79.65
Online exclusive
88.10 
Save 8.45 (10%)
-
Add to cart
Chemical Identifiers
CAS57-13-6
IUPAC Nameurea
Molecular FormulaCH4N2O
InChI KeyXSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILESNC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0-100.5%
Chloride content≤ 5ppm
Residue on ignition≤ 0.01%
Impurity contentSulfate SO₄: 0.001% max
Insoluble material≤ 0.01%
View more
Urea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only