Urea, 99.3+%, Thermo Scientific Chemicals
Urea, 99.3+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Urea, 99.3+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Catalog number 036429.36
also known as 036429-36
Price (USD)/ Each
52.65
Online exclusive
58.00 
Save 5.35 (9%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
52.65
Online exclusive
58.00 
Save 5.35 (9%)
Add to cart
Urea, 99.3+%, Thermo Scientific Chemicals
Catalog number036429.36
Price (USD)/ Each
52.65
Online exclusive
58.00 
Save 5.35 (9%)
-
Add to cart
Chemical Identifiers
CAS57-13-6
IUPAC Nameurea
Molecular FormulaCH4N2O
InChI KeyXSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILESNC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on ignition≤0.01 %
Appearance (Color)White
Assay from Supplier's CofAIron (Fe): ≤0.001 %
Assay from Supplier's CofASulfate (SO₄): ≤0.001 %
Insoluble materialAlcohol Insoluble Substances: ≤0.01%
View more
Urea has important use as a fertilizer and feed supplement. Used for the preparation of plastics and drugs. Urea and its derivatives plays an important role in the cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only