Thermo Scientific Chemicals

Urea, 99.3+%, Thermo Scientific Chemicals

Catalog number: 036429.A4
2.5 kg, Each
Thermo Scientific Chemicals

Urea, 99.3+%, Thermo Scientific Chemicals

Catalog number: 036429.A4
2.5 kg, Each
Quantity
Catalog number: 036429.A4
also known as 036429-A4
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57-13-6
IUPAC Name
urea
Molecular Formula
CH4N2O
InChI Key
XSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILES
NC(N)=O
Residue on ignition
≤0.01 %
Appearance (Color)
White
Assay from Supplier's CofA
Iron (Fe): ≤0.001 %
Assay from Supplier's CofA
Sulfate (SO₄): ≤0.001 %
Insoluble material
Alcohol Insoluble Substances: ≤0.01%

Description

Urea has important use as a fertilizer and feed supplement. Used for the preparation of plastics and drugs. Urea and its derivatives plays an important role in the cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text