Thermo Scientific Chemicals

Sodium tetraphenylborate, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog number: 036430.04
2 g, Each
Thermo Scientific Chemicals

Sodium tetraphenylborate, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog number: 036430.04
2 g, Each
Quantity
Catalog number: 036430.04
also known as 036430-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143-66-8
IUPAC Name
sodium tetraphenylboranuide
Molecular Formula
C24H20BNa
InChI Key
HFSRCEJMTLMDLI-UHFFFAOYSA-N
SMILES
[Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Assay from Supplier's CofA
≥99.5% (Gravimetry)
Loss on Drying
≤0.5% (constant wt. @105?C)
Form
Solid
Clarity
Passes test
Appearance (Color)
White

Description

Sodium tetraphenylborate is used as a precipitating reagent for potassium, as a protecting agent to protect amino acids. It is used in the preparation of N-acylammonium salts. It finds application as a phenyl donor in palladium-catalyzed cross-coupling reactions to prepare arylalkenes and biaryl compounds. It is used in inorganic and organometallic chemistry due to its solubility in nonpolar solvents. Furthermore, it is involved in the isolation of complexes having dinitrogen ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text