Thermo Scientific Chemicals

Trisodium citrate dihydrate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 036439.A3
2 kg, Each
Thermo Scientific Chemicals

Trisodium citrate dihydrate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 036439.A3
2 kg, Each
Quantity
Catalog number: 036439.A3
also known as 036439-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6132-04-3
IUPAC Name
trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
Molecular Formula
C6H9Na3O9
InChI Key
NLJMYIDDQXHKNR-UHFFFAOYSA-K
SMILES
O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Assay (unspecified)
99.0% Na₃C₆H₅O₇.2H₂O min
Impurity content
Fe 5ppm
Impurity content
SO₄ 0.005%
Heavy Metals
(ICP-OES) 5ppm
Impurity content
Cl 0.003%

Description

Trisodium citrate dihydrate, is widely applied in food, beverages and fillers as a buffering, sequestering or an emulsifying agent. It used as an anticoagulant in blood transfusions, osmotic laxative, functional fluids, solvents Cleaning, Furnishing Care Products, Laundry Dishwashing Products and cleaning automobile radiators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text