Thermo Scientific Chemicals

Phenylazoformic acid 2-phenylhydrazide compound with 1,5-Diphenylcarbohydrazide, ACS, Thermo Scientific Chemicals

Catalog number: 036442.09
10 g, Each
Thermo Scientific Chemicals

Phenylazoformic acid 2-phenylhydrazide compound with 1,5-Diphenylcarbohydrazide, ACS, Thermo Scientific Chemicals

Catalog number: 036442.09
10 g, Each
Quantity
Catalog number: 036442.09
also known as 036442-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10329-15-4
IUPAC Name
1,3-bis(phenylamino)urea; 3-(phenylamino)-1-(phenylimino)urea
Molecular Formula
C26H26N8O2
InChI Key
WDJPBIRJSVMEKJ-UHFFFAOYSA-N
SMILES
O=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1.O=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
Residue on ignition
0.1%
Solution Test
in acetone P.T.
Comment
Sensitivity P.T.

Description

Phenylazoformic acid 2-phenylhydrazide compound with 1,5-Diphenylcarbohydrazide is used as an indicator in titrating iron and for the colorimetric determination of chromium and the detection of cadmium, mercury, magnesium, aldehydes, and emetine, a color developing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text