Thermo Scientific Chemicals

Hexamethylenetetramine, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 036462.36
500 g, Each
Thermo Scientific Chemicals

Hexamethylenetetramine, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 036462.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 036462.36
also known as 036462-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-97-0
IUPAC Name
1,3,5,7-tetraazatricyclo[3.3.1.1³,⁷]decane
Molecular Formula
C6H12N4
InChI Key
VKYKSIONXSXAKP-UHFFFAOYSA-N
SMILES
C1N2CN3CN1CN(C2)C3
Appearance (Color)
White
Form
solid
Assay (Titration)
≥99.0% (dried basis)
Residue on ignition
≤0.1%
Loss on Drying
≤2.0%

Description

Hexamethylenetetramine is used in the manufacturing of phenolic resins liquid preparations and molding compounds. It finds applications in in brake and clutch linings, abrasive products, non-woven textiles and fireproof materials. It is also used as a drug taken by mouth to prevent recurrent urinary tract infections. It is used as a solid fuel in association with 1,3,5-trioxane in campers, hobbyists, the military and relief organizations. It acts as a food additive and as a preservative. It is an important reagent in organic synthesis, especially Duff reaction, Sommelet reaction and in the Delepine reaction. Further, it is used in adhesives, coatings, sealants, and leather preservatives. It is utilized as a formaldehyde releaser for photographic film development and as a hardener for epoxy resins of the bisphenol A type.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text