Specifications
Chemical Name or MaterialTris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III)
CAS14319-13-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more