Thermo Scientific Chemicals

Chromium(VI) oxide, ACS, 98+%, Thermo Scientific Chemicals

Catalog number: 036468.22
100 g, Each
Thermo Scientific Chemicals

Chromium(VI) oxide, ACS, 98+%, Thermo Scientific Chemicals

Catalog number: 036468.22
100 g, Each
Quantity
Catalog number: 036468.22
also known as 036468-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1333-82-0
IUPAC Name
trioxochromium
Molecular Formula
CrO3
InChI Key
WGLPBDUCMAPZCE-UHFFFAOYSA-N
SMILES
O=[Cr](=O)=O
Insoluble material
0.01%
Assay (unspecified)
98.0% CrO₃ min
Impurity content
Al 0.02%
Impurity content
Ba 0.01%
Impurity content
Cl 0.005%

Description

Chromium plating, copper stripping, aluminum anodizing, corrosion inhibitor, photography, ceramic glazes, colored glass, oxidizing agent in organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Chromium plating, copper stripping, aluminum anodizing, corrosion inhibitor, photography, ceramic glazes, colored glass, oxidizing agent in organic chemistry
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text