Thermo Scientific Chemicals

Acetic acid-d{4}, 99.5% (Isotopic), Thermo Scientific Chemicals

Catalog number: 036469.09
10 g, Each
Thermo Scientific Chemicals

Acetic acid-d{4}, 99.5% (Isotopic), Thermo Scientific Chemicals

Catalog number: 036469.09
10 g, Each
Quantity
Catalog number: 036469.09
also known as 036469-09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1186-52-3
IUPAC Name
(²H₃)ethan(²H)oic acid
Molecular Formula
C2H4O2
InChI Key
QTBSBXVTEAMEQO-GUEYOVJQSA-N
SMILES
[2H]OC(=O)C([2H])([2H])[2H]
Appearance (Color)
Clear colorless
Assay from Suppliers CofA
≥99.5% ¹H nmr
Form
Liquid
Assay from Suppliers CofA
≥99.5% Isotopic enrichment by ¹H nmr
Water Content (Karl Fischer Titration)
≤ 1000ppm

Description

Acetic acid-d{4} is used as a deuterated, polar and protic solvent, in food preservatives, PH control agent, flavor enhancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text