Thermo Scientific Chemicals

Chloroform-d, 99.8% (Isotopic), contains 1% v/v TMS, Thermo Scientific Chemicals

Catalog number: 036470.22
100 g, Each
Thermo Scientific Chemicals

Chloroform-d, 99.8% (Isotopic), contains 1% v/v TMS, Thermo Scientific Chemicals

Catalog number: 036470.22
100 g, Each
Quantity
Catalog number: 036470.22
also known as 036470-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
865-49-6
IUPAC Name
trichloro(²H)methane
Molecular Formula
CHCl3
InChI Key
HEDRZPFGACZZDS-MICDWDOJSA-N
SMILES
[2H]C(Cl)(Cl)Cl
Proton NMR
>/=99.8% isotopic enrichement
Assay (GC)
Pass/ Fail- >/= 99.5%
Form
liquid
Water Content (Karl Fischer Titration)
100 ppm, max
Concentration (w/v)
0.9 to 1.1 % TMS

Description

Widely used in the organic solvent for the NMR analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text