Thermo Scientific Chemicals

Pyridine-d{5}, 99.5% (Isotopic), Thermo Scientific Chemicals

Catalog number: 036471.04
2 g, Each
Thermo Scientific Chemicals

Pyridine-d{5}, 99.5% (Isotopic), Thermo Scientific Chemicals

Catalog number: 036471.04
2 g, Each
Quantity
Catalog number: 036471.04
also known as 036471-04
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7291-22-7
IUPAC Name
(²H₅)pyridine
Molecular Formula
C5H5N
InChI Key
JUJWROOIHBZHMG-RALIUCGRSA-N
SMILES
[2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]
Form
Liquid
Assay from Supplier's CofA
≥99.5% Isotopic enrichment by ¹H nmr
Appearance (Color)
Clear colorless
Assay from Supplier's CofA
≥99.5% ¹H nmr
Water Content (Karl Fischer Titration)
≤ 500ppm

Description

Pyridine-d{5} is used in the synthesis complexes with cobalt(II) halides, pyridine-d5, a deuterated form of pyridine, ionic liquid matrices (ILMs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text