Thermo Scientific Chemicals

Niobium(V) oxide, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 036478.14
25 g, Each
Thermo Scientific Chemicals

Niobium(V) oxide, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 036478.14
25 g, Each
Quantity
Catalog number: 036478.14
also known as 036478-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1313-96-8
IUPAC Name
[(dioxoniobio)oxy]dioxoniobium
Molecular Formula
Nb2O5
InChI Key
ZKATWMILCYLAPD-UHFFFAOYSA-N
SMILES
O=[Nb](=O)O[Nb](=O)=O
Total Metal Impurities
0.05% max.

Description

Niobium(V) oxide is used for vacuum deposition, as an alternate electrode to lithium metal in advanced fuel cells, Applied in the solid state formation of an unusual cation deficient perovskite. And it is also used in Electrical and Electronic Products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text