Niobium(V) oxide, 99.95% (metals basis), Thermo Scientific Chemicals
Niobium(V) oxide, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium(V) oxide, 99.95% (metals basis), Thermo Scientific Chemicals

For Vacuum Deposition
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 036478.22
also known as 036478-22
Price (USD)/ Each
301.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
301.00
Add to cart
Niobium(V) oxide, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number036478.22
Price (USD)/ Each
301.00
-
Add to cart
Chemical Identifiers
CAS1313-96-8
IUPAC Name[(dioxoniobio)oxy]dioxoniobium
Molecular FormulaNb2O5
InChI KeyZKATWMILCYLAPD-UHFFFAOYSA-N
SMILESO=[Nb](=O)O[Nb](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
Niobium(V) oxide is used for vacuum deposition, as an alternate electrode to lithium metal in advanced fuel cells, Applied in the solid state formation of an unusual cation deficient perovskite. And it is also used in Electrical and Electronic Products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only