Hydriodic acid, ACS, 55-58%, Thermo Scientific Chemicals
Hydriodic acid, ACS, 55-58%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydriodic acid, ACS, 55-58%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
50 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036484.AP
Price (USD)/ Each
270.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
318.00 
Save 47.35 (15%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
270.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
318.00 
Save 47.35 (15%)
Add to cart
Hydriodic acid, ACS, 55-58%, Thermo Scientific Chemicals
Catalog number036484.AP
Price (USD)/ Each
270.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
318.00 
Save 47.35 (15%)
-
Add to cart
Chemical Identifiers
CAS10034-85-2
IUPAC Namehydrogen iodide
Molecular FormulaHI
InChI KeyXMBWDFGMSWQBCA-UHFFFAOYSA-N
SMILESI
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on ignition0.01%
Impurity contentI₂ 0.75%
Impurity contentSO₄ 0.005%
Assay (unspecified)55.0-58.0% HI
Impurity contentChloride and bromide (as Cl) 0.05%
View more
In organic chemistry, it is used as catalyst for the conversion of primary alcohol to alkyl halide. Suitable for use in methoxyl determinations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.