Thermo Scientific Chemicals

Hydriodic acid, ACS, 55-58%, Thermo Scientific Chemicals

Catalog number: 036484.AY
500 mL, 4 x 500mL, Each
Thermo Scientific Chemicals

Hydriodic acid, ACS, 55-58%, Thermo Scientific Chemicals

Catalog number: 036484.AY
500 mL, 4 x 500mL, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 036484.AY
also known as 036484-AY
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10034-85-2
IUPAC Name
hydrogen iodide
Molecular Formula
HI
InChI Key
XMBWDFGMSWQBCA-UHFFFAOYSA-N
SMILES
I
Residue on ignition
0.01%
Impurity content
I₂ 0.75%
Impurity content
SO₄ 0.005%
Assay (unspecified)
55.0-58.0% HI
Impurity content
Chloride and bromide (as Cl) 0.05%

Description

In organic chemistry, it is used as catalyst for the conversion of primary alcohol to alkyl halide. Suitable for use in methoxyl determinations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used as catalyst for conversion of primary alcohol to alkyl halide
  • Suitable for use in methoxyl determinations.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text