Hydriodic acid, ACS, 47%, stab. with 1.5% hypophosphorous acid, Thermo Scientific Chemicals
Hydriodic acid, ACS, 47%, stab. with 1.5% hypophosphorous acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydriodic acid, ACS, 47%, stab. with 1.5% hypophosphorous acid, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036490.AP
also known as 036490-AP
Price (USD)/ Each
229.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
270.00 
Save 40.35 (15%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
229.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
270.00 
Save 40.35 (15%)
Add to cart
Hydriodic acid, ACS, 47%, stab. with 1.5% hypophosphorous acid, Thermo Scientific Chemicals
Catalog number036490.AP
Price (USD)/ Each
229.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
270.00 
Save 40.35 (15%)
-
Add to cart
Chemical Identifiers
CAS10034-85-2
IUPAC Namehydrogen iodide
Molecular FormulaHI
InChI KeyXMBWDFGMSWQBCA-UHFFFAOYSA-N
SMILESI
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentSO₄ 0.005%
CommentStabilizer (H₃PO₂) 1.5%
Impurity contentFe 0.001%
Assay (unspecified)47.0% HI min
Impurity contentChloride and bromide (as Cl) 0.05%
View more
Hydriodic acid is used as a catalyst in the manufacture of acetic acid, as a pharmaceutical intermediate, in various disinfectants and in the preparation of organic and inorganic iodides. It is also used to prepare alkyl iodides from primary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.