Thermo Scientific Chemicals

Acacia, Total ash <4%, Thermo Scientific Chemicals

Catalog number: 036499.36
500 g, Each
Thermo Scientific Chemicals

Acacia, Total ash <4%, Thermo Scientific Chemicals

Catalog number: 036499.36
500 g, Each
Quantity
Catalog number: 036499.36
also known as 036499-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9000-01-5
Form
Flakes or granules or powder
Ash
≤4.0%
Impurity content
≤5ppm Arsenic
Insoluble material
≤0.5% Acid insoluble ash
Insoluble material
≤1.0% Residue

Description

It is used in textiles, ceramics, lithography, cosmetics and pharmaceuticals. In the food industry, acacia is mainly used as confectionery, bakery, dairy, drink, and as a microencapsulating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text