Dichloromethane-d{2}, 99.9% (Isotopic), Thermo Scientific Chemicals
Dichloromethane-d{2}, 99.9% (Isotopic), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloromethane-d{2}, 99.9% (Isotopic), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036500.06
also known as 036500-06
Price (USD)/ Each
130.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
154.00 
Save 23.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
130.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
154.00 
Save 23.35 (15%)
Add to cart
Dichloromethane-d{2}, 99.9% (Isotopic), Thermo Scientific Chemicals
Catalog number036500.06
Price (USD)/ Each
130.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
154.00 
Save 23.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1665-00-5
IUPAC Namedichloro(²H₂)methane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-DICFDUPASA-N
SMILES[2H]C([2H])(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Assay by NMR>/= 99.8% (isotopic)
FormLiquid
In Anion-templated assembly of [2] rotaxanes, the addition of tetrabutylammonium (TBA) halide, dihydrogen phosphate and hydrogen sulfate salts to dichloromethane-d 2 solutions of both derivatives resulted in significant downfield shift perturbations of the respective compounds' aryl b and amide c protons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only