Thermo Scientific Chemicals

p-Toluenesulfonic acid monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: 036506.14
25 g, Each
Thermo Scientific Chemicals

p-Toluenesulfonic acid monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: 036506.14
25 g, Each
Quantity
Catalog number: 036506.14
also known as 036506-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6192-52-5
IUPAC Name
4-methylbenzene-1-sulfonic acid hydrate
Molecular Formula
C7H10O4S
InChI Key
KJIFKLIQANRMOU-UHFFFAOYSA-N
SMILES
O.CC1=CC=C(C=C1)S(O)(=O)=O
Comment
Para content of anhydrous toluenesulfonic acid: ≥99.0%
Impurity content
Sulfuric acid: ≤0.3%
Trace Metal
Iron, Fe: ≤5 ppm (≤0.0005%)
Appearance (Color)
White to pale cream
Form
Crystals or adhering crystals

Description

p-Toluenesulfonic acid monohydrate is used as a catalyst in the synthesis of resveratrol, in oxane derivatives as an antimalarial agent, as substituted piperidine and unsymmetrical benzyl. It acts as a catalyst in the preparation of 1,3,5-trisubstituted pyrazoles derivatives, selenated ketene dithioacetals, triazoloquinazolinone and benzimidazoquinazolinone derivatives. It also serves as an intermediate in the esterification and in reductive amination reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text