Thermo Scientific Chemicals

Sucrose, ACS

Catalog number: 036508.30
250 g, Each
Thermo Scientific Chemicals

Sucrose, ACS

Catalog number: 036508.30
250 g, Each
Quantity
Catalog number: 036508.30
also known as 036508-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57-50-1
IUPAC Name
(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C12H22O11
InChI Key
CZMRCDWAGMRECN-PWPRYFECNA-N
SMILES
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Appearance (Color)
White
Form
Powder or crystalline powder
Chloride content
≤0.005%
Comment
Titrable acid: ≤0.0008 mag/g
Residue on ignition
≤0.01%

Description

Sucrose is widely utilized in medications to carry a more pleasant taste to indigestible chemicals. It is used in many medical dosage forms such as chewable tablets, syrups, tablets, or glues. Sucrose is mainly used in foods nowadays. The fatty acid esters of sucrose show useful application in transdermal therapeutic systems. Sucrose-gradient centrifugation also plays an important role in the selective isolation of nocardia spp. from soil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text